missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tyrosine-protein kinase JAK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Name | Tyrosine-protein kinase JAK1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | JAK1 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031890
|
enQuireBio™
QP6245-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051890
|
enQuireBio™
QP6245-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041890
|
enQuireBio™
QP6245-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061890
|
enQuireBio™
QP6245-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071890
|
enQuireBio™
QP6245-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081890
|
enQuireBio™
QP6245-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Tyrosine-protein kinase JAK1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
JAK1 | |
Recombinant Protein | |
E. coli | |
EEQNPDIVSE KQPTTEVDPT HFEKRFLKRI RDLGEGHFGK VELCRYDPEG DNTGEQVAVK SLKPESGGNH IADLKKEIEI LRNLYHENIV KYKGICMEDG GNGIKLIMEF LPSGSLKEYL PKNKNKINLK QQLKYAIQIC KGMDYLGSRQ YVHRDLAARN VLVESEHQVK IGDFGLTKAI ETDKEYYTVK DDRDSPVFWY APECLIQCKF YIASDVWSFG VTLHELLTYC DSDFSPMALF LKMIGPTHGQ MTVTRLVKTL KEGKRLPCPP NCPDEVYQLM RKCWEFQPSN RTTFQNLIEG FEALL | |
Greater than 90% as determined by SDS-PAGE. |