missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VAMP2 Protein

A cDNA sequence encoding the VAMP2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13916-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985109

Quantity Price
1 114.77 € / 1µg
10 107.33 € / 1µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

VAMP2 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
VAMP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM EDTA, 0.1mM PMSF and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSATAA TVPPAAPAGE GGPPAPPPNL TSNRRLQQTQ AQVDEVVDIM RVNVDKVLER DQKLSELDDR ADALQAGASQ FETSAAKLKR KYWWKNLK
Videos
SDS
Documents
Certificates