missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species Exendin 4 Protein
A cDNA sequence encoding the Exendin 4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 665.00 €
Specifications
Name | Multi Species Exendin 4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Regulates Glucose levels rapidly2. Reduces Insulin resistence 3. Reduces Glucagon4. Reduces HbA1c5. Stimulates beta cell growth which stimulates insulin production. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994348
|
enQuireBio™
QP10593-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974348
|
enQuireBio™
QP10593-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984348
|
enQuireBio™
QP10593-1MG |
1 mg |
665.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Multi Species Exendin 4 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The sequence determined and was found to be His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Regulates Glucose levels rapidly2. Reduces Insulin resistence 3. Reduces Glucagon4. Reduces HbA1c5. Stimulates beta cell growth which stimulates insulin production. | |
Multi Species | |
Untagged | |
The Exendin-4 peptide was lyophilized after extensive dialyses against 20mM PBS and contains 4% mannitol. |