missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species Exendin 4 Protein
A cDNA sequence encoding the Exendin 4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10593-1mg
Additional Details : Weight : 0.01000kg
Specifications
Multi Species Exendin 4 Protein | |
Research Use Only | |
Regulates Glucose levels rapidly2. Reduces Insulin resistence 3. Reduces Glucagon4. Reduces HbA1c5. Stimulates beta cell growth which stimulates insulin production. | |
Multi Species | |
Untagged | |
The Exendin-4 peptide was lyophilized after extensive dialyses against 20mM PBS and contains 4% mannitol. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The sequence determined and was found to be His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |