missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species PRKAR1A Protein
A cDNA sequence encoding the PRKAR1A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13143-3ug
Additional Details : Weight : 0.01000kg
Specifications
5573 | |
3 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MESGSTAASE EARSLRECEL YVQKHNIQAL LKDSIVQLCT ARPERPMAFL REYFERLEKE EAKQIQNLQK AGTRTDSRED EISPPPPNPV VKGRRRRGAI SAEVYTEEDA ASYVRKVIPK DYKTMAALAK AIEKNVLFSH LDDNERSDIF DAMFSVSFIA GETVIQQGDE GDNFYVIDQG ETDVYVNNEW ATSVGEGGSF GELALIYGTP RAATVKAKTN VKLWGIDRDS YRRILMGSTL RKRKMYEEFL SKVSILESLD KWERLTVADA LEPVQFEDGQ KIVVQGEPGD EFFIILEGSA AVLQRRSENE EFVEVGRLGP SDYFGEIALL MNRPRAATVV ARGPLKCVKL DRPRFERVLG PCSDILKRNI QQYNSFVSLS V | |
Greater than 90% as determined by SDS-PAGE. |
Multi Species PRKAR1A Protein | |
Research Use Only | |
PRKAR1A | |
Multi Species | |
Untagged | |
PKA regulatory subunit I a is supplied in 50% glycerol. |