missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species Trp Protein
A cDNA sequence encoding the Trp was constructed and used to recombinantly synthesize the protein.
143.00 € - 433.00 €
Specifications
Name | Multi Species Trp Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Multi Species |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984809
|
enQuireBio™
QP13821-5MG |
5 mg |
143.00 €
5mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964809
|
enQuireBio™
QP13821-10MG |
10 mg |
264.00 €
10mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974809
|
enQuireBio™
QP13821-20MG |
20 mg |
433.00 €
20mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Multi Species Trp Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Multi Species | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |