missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Chaperone protein DnaK Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Chaperone protein DnaK, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6942-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_110122.1 | |
Mycoplasma Chaperone protein DnaK Protein | |
Research Use Only | |
dnaK | |
Recombinant Protein | |
E. coli | |
QGERPMARDN KSLGRFNLGG IQPAPKGKPQ IEITFSLDAN GILNVKAKDL TTQKENSITI SDNGNLSEEE IQKMIRDAEA NKERDNVIRE RIELRNEGES IVSTIKEILQ SPEAKDFPKE EKEKLDKITG GIDAAIKAND YTKLKAEIEN FKKWREEMAK KYNPNGDQGQ PAQ | |
Greater than 90% as determined by SDS-PAGE. |
876838 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |