missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Probable cytosol aminopeptidase, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | NP_110261.1 |
---|---|
Gene ID (Entrez) | 876829 |
Name | Mycoplasma Probable cytosol aminopeptidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036830
|
enQuireBio™
QP6948-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076830
|
enQuireBio™
QP6948-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026830
|
enQuireBio™
QP6948-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056830
|
enQuireBio™
QP6948-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066830
|
enQuireBio™
QP6948-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046830
|
enQuireBio™
QP6948-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_110261.1 | |
Mycoplasma Probable cytosol aminopeptidase Protein | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
876829 | |
Research Use Only | |
pepA | |
Recombinant Protein | |
E. coli | |
MKLNKLFDSK TVVFKKSDKY GKSDCCKTKC VEGQIEFGVK IPTDFPAFNR ALVTFLKTQK NQLNVDLDSF VELYKAENLC YKTALKVVVA SITFCETTPF TMKTEPQKNV EVAVKCDSEH TSLIKEYEVV GNYVNMARQL QDTPSDQLYP EEFVKRFEKA ATGLGVKITV LKQADLIKKK MGLLLGVNKG SEREARLLVI SYNNNKKSSE TLALVGKGIT YDSGGMNIKT GDYMRGMKYD MSGAAIVCST VLALAKNKVK TNVVAVAALT ENLPGPHAQR PDDIQTAYNG KTVEIDNTDA EGRLVLADAI SYAAKDLKAT QIIDVATLTG LMSYILSTTY TGIFSTCDMA WDAFKKAACC AGEPVWRLPM HPDYLKPLES KLADLQNSTS VKGAGSSRAA CFLAEFREGV PLIHCDIAST ASIQDLGQGV LVRTLYERAA QQAKE | |
Greater than 90% as determined by SDS-PAGE. |