missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other 1F8 Chagas Protein
A cDNA sequence encoding the 1F8 Chagas was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | 1F8 Chagas Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995568
|
enQuireBio™
QP10919-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985568
|
enQuireBio™
QP10919-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975568
|
enQuireBio™
QP10919-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1F8 Chagas Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
1F8 Chagas antigen was lyophilized from 20mM Hepes, 300mM NaCl, pH 8.0, and varying 284mM imidazole. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGACGSKGST SDKGLASDKD GKKAKDRKEA WERIRQAIPR EKTAEAKQRR IELFKKFDKN ETGKLCYDEV HSGCLEVLKL DEFTPRVRDI TKRAFDKARA LGSKLENKGS EDFVEFLEFR LMLCYIYDFF ELTVMFDEID ASGNMLVDEE ELKRAVPKLE AWGAKVEDPA ALFKELDKNG TGSVTFDEFA AWASAVKLDA DGDPDNVPES A | |
Protein is >90% pure as determined by SDS PAGE. |