missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other 1F8 Chagas Protein
A cDNA sequence encoding the 1F8 Chagas was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10919-20ug
Additional Details : Weight : 0.01000kg
Specifications
1F8 Chagas Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGACGSKGST SDKGLASDKD GKKAKDRKEA WERIRQAIPR EKTAEAKQRR IELFKKFDKN ETGKLCYDEV HSGCLEVLKL DEFTPRVRDI TKRAFDKARA LGSKLENKGS EDFVEFLEFR LMLCYIYDFF ELTVMFDEID ASGNMLVDEE ELKRAVPKLE AWGAKVEDPA ALFKELDKNG TGSVTFDEFA AWASAVKLDA DGDPDNVPES A | |
Protein is >90% pure as determined by SDS PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
1F8 Chagas antigen was lyophilized from 20mM Hepes, 300mM NaCl, pH 8.0, and varying 284mM imidazole. |