Learn More
enQuireBio™ Recombinant other Avidin Biotin Protein
Recombinant Protein
Brand: enQuireBio™ QP10526-2mg
Additional Details : Weight : 0.01000kg
Specifications
396260 | |
2 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Binding activity of Avidin-Biotin is 10-15units/mg protein. One unit of Avidin activity is defined as the amount of protein which will bind 1microg d-biotin. | |
Other | |
MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE |
Avidin Biotin Protein | |
Research Use Only | |
AVD | |
Recombinant Protein | |
Untagged | |
Salt-free lyophilized powder with no preservative added. |