missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Batroxobin Protein
A cDNA sequence encoding the Batroxobin was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | Batroxobin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Batroxobin’s biological activity was found to be of no less than 500KU/mg. (Klobusitzky Unit). One KU is defined as the amount of enzyme which coagulates standard human plasma incubated at 37C in vitro within 60 ± 20 seconds. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974148
|
enQuireBio™
QP10529-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954148
|
enQuireBio™
QP10529-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964148
|
enQuireBio™
QP10529-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Batroxobin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
VIGGDECDIN EHPFLAFMYY SPRYFCGMTL INQEWVLTAA HCNRRFMRIH LGKHAGSVAN YDEVVRYPKE KFICPNKKKN VITDKDIMLI RLDRPVKNSE HIAPLSLPSN PPSVGSVCRI MGWGAITTSE DTYPDVPHCA NINLFNNTVC REAYNGLPAK TLCAGVLQGG IDTCGGDSGG PLICNGQFQG ILSWGSDPCA EPRKPAFYTK VFDYLPWIQS IIAGNKTATC P | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Batroxobin’s biological activity was found to be of no less than 500KU/mg. (Klobusitzky Unit). One KU is defined as the amount of enzyme which coagulates standard human plasma incubated at 37C in vitro within 60 ± 20 seconds. | |
Other | |
Untagged | |
The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM sodium acetate buffer, pH 7.4. |