missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia BmpA Protein
A cDNA sequence encoding the Borrelia BmpA was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | Borrelia BmpA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966398
|
enQuireBio™
QP11181-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956398
|
enQuireBio™
QP11181-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946398
|
enQuireBio™
QP11181-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Borrelia BmpA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia BmpA (1.3 mg/1ml) is supplied in 20mM HEPES buffer pH 8.0, 6M Urea and 200mM NaCl. |