missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other C. Albicans Enolase Protein

A cDNA sequence encoding the C. Albicans Enolase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11216-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986508

Quantity Price
1 1137.40 € / 100µg
3 1068.13 € / 100µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

C. Albicans Enolase Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
C. Albicans Enolase is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Videos
SDS
Documents
Certificates