missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CST3 k9 Protein
A cDNA sequence encoding the CST3 k9 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CST3 k9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907588
|
enQuireBio™
QP11543-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987578
|
enQuireBio™
QP11543-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997578
|
enQuireBio™
QP11543-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CST3 k9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
K9 Cystatin-Cwas filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS, 50mM NaCl, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS PGAGRRGSRP GAVGGAVDAD VGEEGVQQAL DFAVREYNRA SNDAYHSRAL RVLRARKQVV SGMNYFLEVE IGRTRCTKSQ PNLDNCPFHD QPHLMRKTLC SFQIYTVPWL GKTSLVKSSC QDV | |
Purity is greater than 95% as visualized by SDS-PAGE. |