missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other CST3 k9 Protein

A cDNA sequence encoding the CST3 k9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11543-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987578

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CST3 k9 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
K9 Cystatin-Cwas filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS, 50mM NaCl, pH 7.5.
Purity is greater than 95% as visualized by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS PGAGRRGSRP GAVGGAVDAD VGEEGVQQAL DFAVREYNRA SNDAYHSRAL RVLRARKQVV SGMNYFLEVE IGRTRCTKSQ PNLDNCPFHD QPHLMRKTLC SFQIYTVPWL GKTSLVKSSC QDV
Videos
SDS
Documents
Certificates