missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Cyclophilin A Protein

A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11576-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987688

  • 3401.86 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

947870
Greater than 95% as determined by SDS-PAGE.
Research Use Only
ppiA
Other
His
Cyclophilin A protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
Cyclophilin A Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSAKGDPHV LLTTSAGNIE LELDKQKAPV SVQNFVDYVN SGFYNNTTFH RVIPGFMIQG GGFTEQMQQK KPNPPIKNEA DNGLRNTRGT IAMARTADKD SATSQFFINV ADNAFLDHGQ RDFGYAVFGK VVKGMDVADK ISQVPTHDVG PYQNVPSKPV VILSAKVLP
Videos
SDS
Documents
Certificates