missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cyclosporin A Protein
A cDNA sequence encoding the Cyclosporin A was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | Cyclosporin A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987708
|
enQuireBio™
QP11582-1GRAM |
1 g |
131.00 €
1g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997708
|
enQuireBio™
QP11582-4GRAM |
4 g |
203.00 €
4g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977708
|
enQuireBio™
QP11582-100GRAM |
100 g |
1740.00 €
100g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Cyclosporin A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The Cyclosporin-A was lyophilized from a concentrated (1 mg/ml) solution with no additives. |