missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cyclosporin A Protein
A cDNA sequence encoding the Cyclosporin A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11582-1gram
Additional Details : Weight : 0.01000kg
Specifications
Cyclosporin A Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The Cyclosporin-A was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by RP-HPLC. |