missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DnaJ Protein
A cDNA sequence encoding the DnaJ was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | DnaJ Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957998
|
enQuireBio™
QP11674-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977998
|
enQuireBio™
QP11674-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967998
|
enQuireBio™
QP11674-1MG |
1 mg |
2610.00 €
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DnaJ Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DnaJ contains 25mM Tris-HCl buffer (pH 7.5), 100mM NaCl, 5mM DTT and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAKQDYYEIL GVSKTAEEHE IRKAYKRLAM KYHPDRNQGD KEAEAKFKEI KEAYEVLTDSQKRAAYDQYG HAAFEQGGMG GGGFGGGADF SDIFGDVFGD IFGGGRGRQR AARGADLRYNMELTLEEAVR GVTKEIRIPT LEECDVCHGS GAKPGTQPQT CPTCHGSGQV QMRQGFFAVQQTCPHCQGRG TLIKDPCNKC HGHGRVERSK TLSVKIPAGV DTGDRIRLAG EGEAGEHGAPAGDLYVQVQV KQHPIFEREG NNLYCEVPIN FAMAALGGEI EVPTLDGRVK LKVPGETQTG KLFRMRGKGV KSVRGGAQGD LLCRVVVETP VGLNERQKQL LQELQESFGG PTGEHNSPRSKSFFDGVKKF FDDLTR | |
Greater than 95.0% as determined by SDS-PAGE. |