missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DsbC Protein
A cDNA sequence encoding the DsbC was constructed and used to recombinantly synthesize the protein.
131.00 € - 2030.00 €
Specifications
Gene ID (Entrez) | 947363 |
---|---|
Name | DsbC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | dsbC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908098
|
enQuireBio™
QP11704-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928098
|
enQuireBio™
QP11704-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918098
|
enQuireBio™
QP11704-1MG |
1 mg |
2030.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
947363 | |
Research Use Only | |
dsbC | |
Other | |
Untagged | |
1 mg/ml solution containing 20mM phosphate buffer pH 7.5 and 20mM EDTA. |
DsbC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
DDAAIQQTLA KMGIKSSDIQ PAPVAGMKTV LTNSGVLYIT DDGKHIIQGP MYDVSGTAPVNVTNKMLLKQ LNALEKEMIV YKAPQEKHVI VFTDITCGY CHKLHEQMAD YNALGITVRYLAFPRQGLDS DAEKEMKAIW CAKDKNKAFD DVMAGKSVAP ASCDVDIADH YVLGVQLGVSGTPAVVLSNG TLVPGYQPPK EMKEFLDEHQ KMTSGK | |
Greater than 95.0% as determined by SDS-PAGE. |