missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Enfuvirtide Protein

A cDNA sequence encoding the Enfuvirtide was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11786-500mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988358

Quantity Price
1 1453.80 € / 500mg
3 1367.44 € / 500mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Enfuvirtide Protein
500 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 98.0% as determined by RP-HPLC.
Research Use Only
Recombinant Protein
E. coli
The protein (1 mg/ml) was lyophilized with no additives.
Videos
SDS
Documents
Certificates