missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other ESAT6 Protein
A cDNA sequence encoding the ESAT6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11809-10ug
Additional Details : Weight : 0.01000kg
Specifications
ESAT6 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMTEQQWN FAGIEAAASA IQGNVTSIHS LLDEGKQSLT KLAAAWGGSG SEAYQGVQQK WDATATELNN ALQNLARTIS EAGQAMASTE GNVTGMFAHH HHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
ESAT-6 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |