missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Gliadin Gamma Wheat Protein

A cDNA sequence encoding the Gliadin Gamma Wheat was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11985-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959008

  • 870.00 € / 500µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

Gliadin Gamma Wheat Protein
Research Use Only
Recombinant Protein
E. coli
MKTLLILTILAMAITIGTANIQVDPSGQVQWLQQQLVPQLQQPLSQQPQQTFPQPQQTFPHQPQQQVPQPQQPQQPFLQPQQPFPQQPQQPFPQTQQPQQPFPQQPQQPFPQTQQPQQPFPQQPQQPFPQTQQPQQPFPQLQQPQQPFPQPQQQLPQPQQPQQSFPQQQRPFIQPSLQQQLNCKNILLQQSKPASLVSSLWSIIWPQSDCQVMRQQCCQQLAQIPQQLQCAAIHSVVHSIIMQQQQQQQQQQGIDIFLPLSQHEQVGQGSLVQGQGIIQPQQPAQLEAIRSLVLQTLPSMCNVYVPPECSIMRAPFASIVAGIGGQHHHHHH
Protein is >90% pure.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Gliadin Gamma protein solution (1 mg/ml) in 10mM Tris-HCl pH 7.2.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers