missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GST Protein
A cDNA sequence encoding the GST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10656-50ug
Additional Details : Weight : 0.01000kg
Specifications
GST Protein | |
Research Use Only | |
Specific activity is >20 units/mg. A unit is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C. | |
Other | |
Untagged | |
GST supplied in Phosphate Buffered Saline pH 7.4. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSPILGYWKI KGLVQPTRLL LEYLEEKYEE HLYERDEGDK WRNKKFELGL EFPNLPYYID GDVKLTQSMA IIRYIADKHN MLGGCPKERA EISMLEGAVL DIRYGVSRIA YSKDFETLKV DFLSKLPEML KMFEDRLCHK TYLNGDHVTH PDFMLYDALD VVLYMDPMCL DAFPKLVCFK KRIEAIPQID KYLKSSKYIA WPLQGWQATF GGGDHPPKSD LVPR | |
Greater than 95% as determined by SDS-PAGE. |