missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HBcAg 1-149 a. a. Protein
A cDNA sequence encoding the HBcAg 1-149 a. a. was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12119-500ug
Additional Details : Weight : 0.01000kg
Specifications
HBcAg 1-149 a. a. Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline, pH 7.4. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |