missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other hchA Protein
A cDNA sequence encoding the hchA was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | hchA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15999478
|
enQuireBio™
QP12135-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989478
|
enQuireBio™
QP12135-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979478
|
enQuireBio™
QP12135-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
hchA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The hchA contains (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTVQTSKNPQ VDIAEDNAFF PSEYSLSQYT SPVSDLDGVD YPKPYRGKHK ILVIAADERY LPTDNGKLFS TGNHPIETLL PLYHLHAAGF EFEVATISGL MTKFEYWAMP HKDEKVMPFF EQHKSLFRNP KKLADVVASL NADSEYAAIF VPGGHGALIG LPESQDVAAA LQWAIKNDRF VISLCHGPAA FLALRHGDNP LNGYSICAFP DAADKQTPEI GYMPGHLTWY FGEELKKMGM NIINDDITGR VHKDRKLLTG DSPFAANALG KLAAQEMLAA YAG | |
Greater than 95.0% as determined by SDS-PAGE. |