missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other hchA Protein

A cDNA sequence encoding the hchA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12135-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989478

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

hchA Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
The hchA contains (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MTVQTSKNPQ VDIAEDNAFF PSEYSLSQYT SPVSDLDGVD YPKPYRGKHK ILVIAADERY LPTDNGKLFS TGNHPIETLL PLYHLHAAGF EFEVATISGL MTKFEYWAMP HKDEKVMPFF EQHKSLFRNP KKLADVVASL NADSEYAAIF VPGGHGALIG LPESQDVAAA LQWAIKNDRF VISLCHGPAA FLALRHGDNP LNGYSICAFP DAADKQTPEI GYMPGHLTWY FGEELKKMGM NIINDDITGR VHKDRKLLTG DSPFAANALG KLAAQEMLAA YAG
Videos
SDS
Documents
Certificates