missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HDV 167 a. a Protein
A cDNA sequence encoding the HDV 167 a. a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12213-100ug
Additional Details : Weight : 0.01000kg
Specifications
HDV 167 a. a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 10mM CBB, pH 10, 100mM NaCl and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |