missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Hirudin Protein
A cDNA sequence encoding the Hirudin was constructed and used to recombinantly synthesize the protein.
131.00 € - 5920.00 €
Specifications
Name | Hirudin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity was found to be >14,000ATU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904588
|
enQuireBio™
QP10666-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984578
|
enQuireBio™
QP10666-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994578
|
enQuireBio™
QP10666-1MG |
1 mg |
5920.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hirudin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
Each mg of protein was lyophilized from a sterile solution containing 20mM PBS pH 7 and 2% mannitol. |
Research Use Only | |
The specific activity was found to be >14,000ATU/mg. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |