missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HPSE WB Protein
Recombinant Protein
665.00 € - 1970.00 €
Specifications
Gene ID (Entrez) | 10855 |
---|---|
Name | HPSE WB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HPSE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939998
|
enQuireBio™
QP12305-100NG |
100 ng |
665.00 €
0.10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949998
|
enQuireBio™
QP12305-200NG |
200 ng |
1305.00 €
0.20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959998
|
enQuireBio™
QP12305-300NG |
300 ng |
1970.00 €
0.20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10855 | |
Research Use Only | |
HPSE | |
Other | |
Untagged | |
Concentration: 1ug /ml Content: 100 ngBuffer: LDS-PAGE buffer[140 mM Tris buffer pH 8.5, 10% Glycerol, 2% LDS, 0.015% EDTA, 1.88% (v/v) of 1% Serva Blue G250 and 0.625% (v/v) of 1% Phenol red]. |
HPSE WB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MLLRSKPALP PPLMLLLLGP LGPLSPGALP RPAQAQDVVD LDFFTQEPLH LVSPSFLSVT IDANLATDPR FLILLGSPKL RTLARGLSPA YLRFGGTKTD FLIFDPKKES TFEERSYWQS QVNQDICKYG SIPPDVEEKL RLEWPYQEQL LLREHYQKKF KNSTYSRSSV DVLYTFANCS GLDLIFGLNA LLRTADLQWN SSNAQLLLDY CSSKGYNISW ELGNEPNSFL KKADIFINGS QLGEDFIQLH KLLRKSTFKN AKLYGPDVGQ PRRKTAKMLK SFLKAGGEVI DSVTWHHYYL NGRTATKEDF LNPDVLDIFI SSVQKVFQVV ESTRPGKKVW LGETSSAYGG GAPLLSDTFA AGFMWLDKLG LSARMGIEVV MRQVFFGAGN YHLVDENFDP LPDYWLSLLF KKLVGTKVLM ASVQGSKRRK LRVYLHCTNT DNPRYKEGDL TLYAINLHNV TKYLRLPYPF SNKQVDKYLL RPLGPHGLLS KSVQLNGLTL KMVDDQTLPP LMEKPLRPGS SLGLPAFSYS FFVIRNAKVA ACI |