missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HRP Protein
A cDNA sequence encoding the HRP was constructed and used to recombinantly synthesize the protein.
131.00 € - 2900.00 €
Specifications
Name | HRP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Greater than 268 U/mg (25C, guaiacol as the hydrogen donor, pH 7 and H2O2 as substrates). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984588
|
enQuireBio™
QP10669-20MG |
20 mg |
131.00 €
20mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994588
|
enQuireBio™
QP10669-60MG |
60 mg |
203.00 €
60mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974588
|
enQuireBio™
QP10669-1GRAM |
1 g |
2900.00 €
1g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HRP Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Non-species Specific |
Research Use Only | |
Greater than 268 U/mg (25C, guaiacol as the hydrogen donor, pH 7 and H2O2 as substrates). | |
Other | |
Untagged |