missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HSP104 Saccharomyces Protein
A cDNA sequence encoding the HSP104 Saccharomyces was constructed and used to recombinantly synthesize the protein.
131.00 € - 2030.00 €
Specifications
Name | HSP104 Saccharomyces Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990059
|
enQuireBio™
QP12325-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910069
|
enQuireBio™
QP12325-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900069
|
enQuireBio™
QP12325-1MG |
1 mg |
2030.00 €
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSP104 Saccharomyces Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The HSP-104 protein solution contains 20mM Tris-HCl, pH 7.4,100mM NaCl, 2mM EDTA and 5% Glycerol. |
Research Use Only | |
Recombinant Protein | |
S. cerevisiae | |
MNDQTQFTER ALTILTLAQK LASDHQHPQL QPIHILAAFI ETPEDGSVPY LQNLIEKGRY DYDLFKKVVN RNLVRIPQQQ PAPAEITPSY ALGKVLQDAA KIQKQQKDSF IAQDHILFAL FNDSSIQQIF KEAQVDIEAI KQQALELRGN TRIDSRGADT NTPLEYLSKY AIDMTEQARQ GKLDPVIGRE EEIRSTIRVL ARRIKSNPCL IGEPGIGKTA IIEGVAQRII DDDVPTILQG AKLFSLDLAA LTAGAKYKGD FEERFKGVLK EIEESKTLIV LFIDEIHMLM GNGKDDAANI LKPALSRGQL KVIGATTNNE YRSIVEKDGA FERRFQKIEV AEPSVRQTVA ILRGLQPKYE IHHGVRILDS ALVTAAQLAK RYLPYRRLPD SALDLVDISC AGVAVARDSK PEELDSKERQ LQLIQVEIKA LERDEDADST TKDRLKLARQ KEASLQEELE PLRQRYNEEK HGHEELTQAK KKLDELENKA LDAERRYDTA TAADLRYFAI PDIKKQIEKL EDQVAEEERR AGANSMIQNV VDSDTISETA ARLTGIPVKK LSESENEKLI HMERDLSSEV VGQMDAIKAV SNAVRLSRSG LANPRQPASF LFLGLSGSGK TELAKKVAGF LFNDEDMMIR VDCSELSEKY AVSKLLGTTA GYVGYDEGGF LTNQLQYKPY SVLLFDEVEK AHPDVLTVML QMLDDGRITS GQGKTIDCSN CIVIMTSNLG AEFINSQQGS KIQESTKNLV MGAVRQHFRP EFLNRISSIV IFNKLSRKAI HKIVDIRLKE IEERFEQNDK HYKLNLTQEA KDFLAKYGYS DDMGARPLNR LIQNEILNKL ALRILKNEIK DKETVNVVLK KGKSRDENVP EEAEECLEVL PNHEATIGAD TLGDDDNEDS MEIDDDLD | |
Greater than 90.0% as determined by SDS-PAGE. |