missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HYAL1 Protein

A cDNA sequence encoding the HYAL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12361-4500IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990169

Quantity Price
1 171.64 € / 4500 units
10 161.27 € / 4500 units
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

HYAL1 Protein
Research Use Only
Recombinant Protein
Bovine
The enzyme was lyophilized 1xPBS and 2% sucrose.
4500 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Videos
SDS
Documents
Certificates