missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other LDHB Protein
A cDNA sequence encoding the LDHB was constructed and used to recombinantly synthesize the protein.
131.00 € - 1425.00 €
Specifications
Name | LDHB Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity was found to be 258 U/mg protein. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945038
|
enQuireBio™
QP10744-1MG |
1 mg |
131.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965038
|
enQuireBio™
QP10744-3MG |
3 mg |
203.00 €
3mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955038
|
enQuireBio™
QP10744-30MG |
30 mg |
1425.00 €
30mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LDHB Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with 0.1 mg potassium phosphate. |
Research Use Only | |
The specific activity was found to be 258 U/mg protein. | |
Other | |
Untagged | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |