missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MIP2 Viral Protein
A cDNA sequence encoding the MIP2 Viral was constructed and used to recombinantly synthesize the protein.
131.00 € - 1970.00 €
Specifications
Name | MIP2 Viral Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Determined by the inhibitory effect on monocyte migration response to human MIP1A using a concentration range of 1microg-10microg/ml of viral MIP2 which will inhibit 25ng/ml of human MIP1A. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985168
|
enQuireBio™
QP10788-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905178
|
enQuireBio™
QP10788-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995168
|
enQuireBio™
QP10788-1MG |
1 mg |
1970.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MIP2 Viral Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
LGASWHRPDK CCLGYQKRPL PQVLLSSWYP TSQLCSKPGV IFLTKRGRQVCADKSKDWVK KLMQQLPVTA | |
Greater than 97.0% as determined by RP-HPLC & SDS-PAGE. |
Research Use Only | |
Determined by the inhibitory effect on monocyte migration response to human MIP1A using a concentration range of 1microg-10microg/ml of viral MIP2 which will inhibit 25ng/ml of human MIP1A. | |
Other | |
Untagged | |
Lyophilized from 1 mg/ml solution containing 20mM Phospahte Buffer, pH 7.4 % 0.15M NaCl. |