missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MSRA Protein
A cDNA sequence encoding the MSRA was constructed and used to recombinantly synthesize the protein.
131.00 € - 1595.00 €
Specifications
Name | MSRA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911399
|
enQuireBio™
QP12742-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931399
|
enQuireBio™
QP12742-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921399
|
enQuireBio™
QP12742-50UG |
50 μg |
1595.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MSRA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
MSRA protein solution (0.5 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSLFDKKHLV SPADALPGRN TPMPVATLHA VNGHSMTNVP DGMEIAIFAM GCFWGVERLF WQLPGVYSTA AGYTGGYTPN PTYREVCSGD TGHAEAVRIV YDPSVISYEQ LLQVFWENHD PAQGMRQGND HGTQYRSAIY PLTPEQDAAA RASLERFQAA MLAADDDRHI TTEIANATPF YYAEDDHQQY LHKNPYGYCG IGGIGVCLPP EA | |
Greater than 90.0% as determined by SDS-PAGE. |