missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MT II Protein
A cDNA sequence encoding the MT II was constructed and used to recombinantly synthesize the protein.
433.00 € - 2900.00 €
Specifications
Name | MT II Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941409
|
enQuireBio™
QP12746-20MG |
20 mg |
433.00 €
20mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931409
|
enQuireBio™
QP12746-100MG |
100 mg |
1015.00 €
100mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951409
|
enQuireBio™
QP12746-500MG |
500 mg |
2900.00 €
500mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MT II Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |