missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PCC Protein
A cDNA sequence encoding the PCC was constructed and used to recombinantly synthesize the protein.
131.00 € - 1015.00 €
Specifications
Name | PCC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932119
|
enQuireBio™
QP12973-10IU |
10 IU |
131.00 €
10 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942119
|
enQuireBio™
QP12973-300IU |
300 IU |
1015.00 €
300 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952119
|
enQuireBio™
QP12973-50IU |
50 IU |
203.00 €
50 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PCC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized Powder from 0.1M sodium chloride and 10mM sodium citrate buffer. |