missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Phospholipase A2 Protein
A cDNA sequence encoding the Phospholipase A2 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1450.00 €
Specifications
Gene ID (Entrez) | 406141 |
---|---|
Name | Phospholipase A2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Pla2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932339
|
enQuireBio™
QP13045-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952339
|
enQuireBio™
QP13045-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942339
|
enQuireBio™
QP13045-1MG |
1 mg |
1450.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
406141 | |
Research Use Only | |
Pla2 | |
Other | |
Untagged | |
20mM Tris-HCl, pH 7.2, 1.5M urea and 50% glycerol. |
Phospholipase A2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MQVVLGSLFL LLLSTSHGWQ IRDRIGDNEL EERIIYPGTL WCGHGNKSSG PNELGRFKHT DACCRTHDMC PDVMSAGESK HGLTNTASHT RLSCDCDDKF YDCLKNSADT ISSYFVGKMY FNLIDTKCYK LEHPVTGCGE RTEGRCLHYT VDKSKPKVYQ WFDLRKY | |
Protein is >90% pure as determined by 10% SDS-PAGE. |