missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Placental Lactogen Caprine Protein

A cDNA sequence encoding the Placental Lactogen Caprine was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10828-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985288

Quantity Price
1 633.84 € / 1mg
5 594.29 € / 1mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Placental Lactogen Caprine Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3.
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Placental Lactogen Caprine is biologically active as evidenced by inducing proliferation of Nb2 cells.
Other
Untagged
Videos
SDS
Documents
Certificates