missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other SecB Protein

A cDNA sequence encoding the SecB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13448-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933599

  • 120.00 € / 5µg
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SecB Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The SecB protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8 and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSEQNNTEMT FQIQRIYTKD ISFEAPNAPH VFQKDWQPEV KLDLDTASSQ LADDVYEVVL RVTVTASLGE ETAFLCEVQQ GGIFSIAGIE GTQMAHCLGA YCPNILFPYA RECITSMVSR GTFPQLNLAP VNFDALFMNY LQQQAGEGTE EHQDA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers