missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Staphylokinase Protein
A cDNA sequence encoding the Staphylokinase was constructed and used to recombinantly synthesize the protein.
131.00 € - 1040.00 €
Specifications
Name | Staphylokinase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The biological activity measured by the ability of fibrin lysis in agarose plate was found to be 50,000IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15905428
|
enQuireBio™
QP10870-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985418
|
enQuireBio™
QP10870-100UG |
100 μg |
211.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995418
|
enQuireBio™
QP10870-1MG |
1 mg |
1040.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Staphylokinase Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLKRSLLFLT VLLLLFSFSS ITNEVSASSS FDKGKYKKGD DASYFEPTGP YLMVNVTGVD GKGNELLSPH YVEFPIKPGT TLTKEKIEYY VEWALDATAY KEFRVVELDP SAKIEVTYYD KNKKKEETKS FPITEKGFVV PDLSEHIKNP GFNLITKVVI EKK | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The biological activity measured by the ability of fibrin lysis in agarose plate was found to be 50,000IU/mg. | |
Other | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |