missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Staphylokinase Protein
A cDNA sequence encoding the Staphylokinase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10870-100ug
Additional Details : Weight : 0.01000kg
Especificaciones
Staphylokinase Protein | |
Research Use Only | |
The biological activity measured by the ability of fibrin lysis in agarose plate was found to be 50,000IU/mg. | |
Other | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLKRSLLFLT VLLLLFSFSS ITNEVSASSS FDKGKYKKGD DASYFEPTGP YLMVNVTGVD GKGNELLSPH YVEFPIKPGT TLTKEKIEYY VEWALDATAY KEFRVVELDP SAKIEVTYYD KNKKKEETKS FPITEKGFVV PDLSEHIKNP GFNLITKVVI EKK | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |