missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other T. pallidum p15 (Partial) Protein

A cDNA sequence encoding the T. pallidum p15 (Partial) was constructed and used to recombinantly synthesize the protein.

228.00 € - 1740.00 €

Specifications

Name T. pallidum p15 (Partial) Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15954329
View Documents Promotion Details
enQuireBio™
QP13670-GST-100UG
100 μg
228.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15974329
View Documents Promotion Details
enQuireBio™
QP13670-GST-500UG
500 μg
870.00 €
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15964329
View Documents Promotion Details
enQuireBio™
QP13670-GST-1MG
1 mg
1740.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

T. pallidum p15 (Partial) Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
GST
Treponema Pallidum protein is >95% pure as determined by SDS- PAGE.
Research Use Only
Recombinant Protein
E. coli
70mM Tris-HCl pH 8.0, 50mM NaCl, 50% Glycerol, 1.5 M Urea.
Videos
SDS
Documents

Documents

Product Certifications