missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p41 Mosaic Protein
A cDNA sequence encoding the T. pallidum p41 Mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13674-1mg
Additional Details : Weight : 0.01000kg
Specifications
T. pallidum p41 Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 60mM NaCl and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
GST | |
Treponema Pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). |