missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Thymosin ?4 Protein
A cDNA sequence encoding the Thymosin ?4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1440.00 €
Specifications
Name | Thymosin ?4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944569
|
enQuireBio™
QP13746-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924569
|
enQuireBio™
QP13746-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934569
|
enQuireBio™
QP13746-1MG |
1 mg |
1440.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Thymosin ?4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Lyophilized from a 0.2um filtered concentrated solution in 1xPBS, pH 7.4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |