missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other UPP1ella Protein
A cDNA sequence encoding the UPP1ella was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13901-50ug
Additional Details : Weight : 0.01000kg
Specifications
UPP1ella Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSKSDVFHLG LTKNDLQGAQ LAIVPGDPER VEKIAALMDK PVKLASHREF TSWRAELDGK AVIVCSTGIG GPSTSIAVEE LAQLGIRTFL RIGTTGAIQP HINVGDVLVT TASVRLDGAS LHFAPMEFPA VADFACTTAL VEAAKSIGAT THVGVTASSD TFYPGQERYD TYSGRVVRRF KGSMEEWQAM GVMNYEMESA TLLTMCASQG LRAGMVAGVI VNRTQQEIPN AETMKQTESH AVKIVVEAAR RLL |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
30 U/mg protein. | |
Other | |
Untagged | |
The UPase was lyophilized from 1 mg/ml solution containing 25mM Tris-HCl, pH 8.0, 0.15M NaCl. |