missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine GHBP Protein
A cDNA sequence encoding the GHBP was constructed and used to recombinantly synthesize the protein.
131.00 € - 4880.00 €
Specifications
Name | GHBP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Ovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15998948
|
enQuireBio™
QP11966-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988948
|
enQuireBio™
QP11966-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978948
|
enQuireBio™
QP11966-1MG |
1 mg |
4880.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GHBP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ovine | |
Untagged | |
GHBP was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDLWQLLLTL AVAGSSDAFS GSEATPAFFV RASQSLQILY PGLETNSSGN LKFTKCRSPE LETFSCHWTD GANHSLQSPG SVQMFYIRRD IQEWKECPDY VSAGENSCYF NSSYTSVWTP YCIKLTSNGG IVDHKCFSVE DIVQPDPPVG LNWTLLNISL TEIHADILVK WEPPPNTDVK MGWIILEYEL HYKELNETQW KMMDPLLVTS VPMYSLRLDK EYEVRVRTRQ RNTEKYGKFS EVLLITFPQM NPSACEEDFQ FPWFLIIIFG ILGLTVTLFL LIFSKQQRIK MLILPPVPVP KIKGIDPDLL KEGKLEEVNT ILAIHDNYKH EFYNDDSWVE FIELDIDDPD EKTEGSDTDR LLSNDHEKSL SIFGAKDDDS GRTSCYEPDI LETDFHVSDM CDGTSEVAQP QRLKGEADIL CLDQKNQNNS PSNDAAPASQ QPSVILVEEN KPRPLPIGGT ESTHQAVHTQ LSNPSSLANI DFYAQVSDIT PAGNVVLSPG QKNKTGNPQC DTHPEVVTPS QADFIVDSAY FCEVDAKKYI ALAPDVEAES HIEPSFNQED IYITTESLTT TAGRSGTAEN VPSSEIPVPD YTSIHIVQSP QGLVLNATAL PLPDKEFLSS CGYVSTDQLN KIMP | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |