missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine GHBP Protein

A cDNA sequence encoding the GHBP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11966-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988948

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

GHBP Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Ovine
Untagged
GHBP was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MDLWQLLLTL AVAGSSDAFS GSEATPAFFV RASQSLQILY PGLETNSSGN LKFTKCRSPE LETFSCHWTD GANHSLQSPG SVQMFYIRRD IQEWKECPDY VSAGENSCYF NSSYTSVWTP YCIKLTSNGG IVDHKCFSVE DIVQPDPPVG LNWTLLNISL TEIHADILVK WEPPPNTDVK MGWIILEYEL HYKELNETQW KMMDPLLVTS VPMYSLRLDK EYEVRVRTRQ RNTEKYGKFS EVLLITFPQM NPSACEEDFQ FPWFLIIIFG ILGLTVTLFL LIFSKQQRIK MLILPPVPVP KIKGIDPDLL KEGKLEEVNT ILAIHDNYKH EFYNDDSWVE FIELDIDDPD EKTEGSDTDR LLSNDHEKSL SIFGAKDDDS GRTSCYEPDI LETDFHVSDM CDGTSEVAQP QRLKGEADIL CLDQKNQNNS PSNDAAPASQ QPSVILVEEN KPRPLPIGGT ESTHQAVHTQ LSNPSSLANI DFYAQVSDIT PAGNVVLSPG QKNKTGNPQC DTHPEVVTPS QADFIVDSAY FCEVDAKKYI ALAPDVEAES HIEPSFNQED IYITTESLTT TAGRSGTAEN VPSSEIPVPD YTSIHIVQSP QGLVLNATAL PLPDKEFLSS CGYVSTDQLN KIMP
Videos
SDS
Documents
Certificates